Multi-gas detector, GasAlert MicroClip
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
DM6000
SIMEDM6000EA 869 EUR
SIMEDM6000
Multi-gas detector, GasAlert MicroClip
Gasdetectiesystemen
The GasAlert MicroClip offers simultaneous monitoring of two gasses (O₂, LEL) in a rugged compact case for confined areas.
  • Hazardous materials (HAZMAT) response and other applications requiring gas monitoring
  • One-button operation with automatic calibration
  • Can record and transmit up to ten gas alarm events
Leveringsinformatie: Supplied with sensors, lithium battery, stainless steel belt clip, calibration hose/cap and manual.