Temperature indicating stickers, Series 1
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TC9121 TC9116 TC9110 TC9104 TC9099 TC9093 TC9088 TC9082 TC9071 TC9065 TC9060 TC9049 TC9044 TC9040 TC9054 TC9254 TC9241 TC9224 TC9216 TC9204 TC9199 TC9182 TC9177 TC9171 TC9160 TC9149 TC9143 TC9132 TC9077
SIMETC9121EA 38.7 EUR
SIMETC9121 SIMETC9116 SIMETC9110 SIMETC9104 SIMETC9099 SIMETC9093 SIMETC9088 SIMETC9082 SIMETC9071 SIMETC9065 SIMETC9060 SIMETC9049 SIMETC9044 SIMETC9040 SIMETC9054 SIMETC9254 SIMETC9241 SIMETC9224 SIMETC9216 SIMETC9204 SIMETC9199 SIMETC9182 SIMETC9177 SIMETC9171 SIMETC9160 SIMETC9149 SIMETC9143 SIMETC9132 SIMETC9077
Temperature indicating stickers, Series 1
Temperatuurindicatiestrips
These single temperature indicating stickers are designed for use in electronics, food and pharmaceutical industries, and also in the oven and engine construction businesses. They are particularly useful for indicating temperatures of moving or inaccessible parts.
  • Self-adhesive foils with to indicate whether the indicated temperature has been exceeded
  • Indicator changes colour when marked temperature is reached and the change is irreversible
Leveringsinformatie: Series 1 contains 50 stickers per sheet.