Thermometer with surface probe, TQC Thermapen Fast Response
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
TE0035
SIMETE0035EA 176 EUR
SIMETE0035
Thermometer with surface probe, TQC Thermapen Fast Response
Thermometers Digitale thermometers
The TQC Sheen Thermapen Fast Response thermometer with surface probe is designed for precise temperature measurement in viscosity testing applications within the industrial sector.
  • Pivoting probe and swivel head ensure optimal surface contact
  • User selectable resolutions of 0,1 or 1 °C
  • Convenient one-handed operation, activates automatically when opened
  • Biomaster protection inhibits bacterial growth
  • Splashproof body designed for industrial use
  • Complies with European Standard EN 13485
  • Free protective pouch included
  • Optional silicone rubber boot for high-
More Product Information