Viscosity flow cups
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VF2196 VF2197 VF2198 VF2195
SIMEVF2196EA 183 EUR
SIMEVF2196 SIMEVF2197 SIMEVF2198 SIMEVF2195
Viscosity flow cups
Viscosimeters Flow-cup viscometers
These viscosity flow cups are titanium anodised aluminum viscosity cups.
Suitable to be used with stand, to measure the viscosity of lacquers, paint and other liquids.
Certificaten: AFNOR