Viscosity flow cup
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VF2199
SIMEVF2199EA 198 EUR
SIMEVF2199
Viscosity flow cup
Viscosimeters Flow-cup viscometers
Lory viscosity cup designed for quick measurements on location or during production processes.
  • Serial number, easy to clean for quick testing
  • Brass zinc plate cup with steel leg (height of 170 mm)
  • Bottom of this cup provided with a small gap as well as a needle point
The flow time is determined when the product flows out of the small gap and as soon as the needle point appears.