Test panels
Leverancier: Industrial Physics
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
VF8521 VF8520 VF8533 VF8532 VF8531 VF8530 VF8527 VF8526 VF8525 VF8522 VF8523 VF8519 VF8518 VF8517 VF8516 VF8515 VF8513 VF8512 VF8511 VF8510 VF8524 VF8529
SIMEVF8521EA 813 EUR
SIMEVF8521 SIMEVF8520 SIMEVF8533 SIMEVF8532 SIMEVF8531 SIMEVF8530 SIMEVF8527 SIMEVF8526 SIMEVF8525 SIMEVF8522 SIMEVF8523 SIMEVF8519 SIMEVF8518 SIMEVF8517 SIMEVF8516 SIMEVF8515 SIMEVF8513 SIMEVF8512 SIMEVF8511 SIMEVF8510 SIMEVF8524 SIMEVF8529
Test panels
Coatinglaag testers
Test panels are used to enhance reproducibility of physical and chemical tests. Each panel is equipped with a hole for hanging and handling.
  • Consistent and reproducible quality with sophisticated finishing: No burrs or sharp edges
  • Physical testing: Adhesion, elongation, impact resistance and elasticity
  • Appearance testing of coatings (gloss, colour, texture)
Order Now
Specificaties
Environmentally Preferable