Sample tubes

Leverancier: Simport Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C572-1 C572-2
SIMPC572-1EA 368 EUR
SIMPC572-1 SIMPC572-2
Sample tubes
Monsternamepotjes Containers voor algemeen gebruik
These tubes are chemically resistant and shatterproof, they are supplied with a leakproof screw cap, particularly important when transporting hazardous material.

  • Tube made of PP
  • Cap made of PE
  • Self-standing base with graduation
  • Leakproof screw cap
  • Available sterile and non-sterile
  • Functional temperature range: –90 to +121 °C

Conical bottom tubes with self-standing base graduated from 10 to 30 ml and 10 to 50 ml respectively in 5 ml increments. Ideal for transport and storage of urine, sputum and most liquids or particulate samples. Molded-inMore Product Information
Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR