Sample tubes
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
T501-1AT T501-5T T501-4AT T501-2AT T501-3AT T501-2T T501-4T T501-5AT T501-5TPR T501-5ATPR T501-4ATPR T501-1ATPR T501-2TPR T501-2ATPR T501-3ATPR T501-4TPR
212-9336EA 203 EUR
212-9336 212-9343 212-9340 212-9337 212-9339 212-9338 212-9341 212-9342 SIMPT501-5TPR SIMPT501-5ATPR SIMPT501-4ATPR SIMPT501-1ATPR SIMPT501-2TPR SIMPT501-2ATPR SIMPT501-3ATPR SIMPT501-4TPR
Sample tubes
Buisjes Reageerbuisjes
PP, non toxic, translucent.
  • Very good chemical resistance
  • Smooth and uniform inner surface
  • Tubes can be autoclaved (121° C) in upright position with caps loosened
  • Round bottom tubes only can be centrifuged up to 17000×g
Designed for the storage and transportation of biological material.
*For size only, cap is included.