Screw caps for T501 sample tubes
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
T501N T502N T501Y T501B T501DG T501L T502Y T502L T501R T502R T501W T502B T502GY T502W T501GY T501O T502DG
212-9344EA 105 EUR
212-9344 212-9345 SIMPT501Y SIMPT501B SIMPT501DG SIMPT501L SIMPT502Y SIMPT502L SIMPT501R SIMPT502R SIMPT501W SIMPT502B SIMPT502GY SIMPT502W SIMPT501GY SIMPT501O SIMPT502DG
Screw caps for T501 sample tubes
Sluitingen Reageerbuissluitingen
Caps are available either with or without a silicone washer between the cap and the tube to ensure a positive leakproof seal at all temperatures.
  • Made of polypropylene
  • Available with or without silicone washer
  • Long skirt and super fast thread design for easy seal
  • Autoclavable
As for tubes, these closures are also made of polypropylene, providing the same coefficient of expansion for both, which further enhances the leakproof qualities of the vials at changing temperatures. The cap features a long skirt and a super-fast thread design that allows it toMore Product Information
Order Now
Specificaties
Environmentally Preferable