Storage Box for Sample Tubes
Leverancier: Simport Scientific
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
T514-2100B T514-581Y T514-225B T514-225G T514-225P T514-225Y T514-2100G T514-2100P T514-2100Y T514-281B T514-281G T514-281P T514-281Y T514-581B T514-581G T514-581P
SIMPT514-2100BEA 231 EUR
SIMPT514-2100B SIMPT514-581Y SIMPT514-225B SIMPT514-225G SIMPT514-225P SIMPT514-225Y SIMPT514-2100G SIMPT514-2100P SIMPT514-2100Y SIMPT514-281B SIMPT514-281G SIMPT514-281P SIMPT514-281Y SIMPT514-581B SIMPT514-581G SIMPT514-581P
Storage Box for Sample Tubes
Dozen Bakken voor buisjes
Cover and base made of polystyrene.
  • Sturdy
  • Stackable
  • Wide variety of economical storage boxes
Printed with a series of numbered squares and the surface accepts the writing with markers by facilitating inventory control.
These storage boxes are designed with a transparent cover that allows the user to see the contents of the box and is keyed to the base to prevent misalignment.