Cannulas for micro syringes, Dosys™
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
370.0840 370.1250 370.22100 370.22150
SOCO370.0840EA 62 EUR
SOCO370.0840 SOCO370.1250 SOCO370.22100 SOCO370.22150
Cannulas for micro syringes, Dosys™
Canules
Blunt end cannulas provide precise distribution with Luer nozzle microdispensers.