Single channel micro pipettes, mechanical, fixed volume, Acura® manual 815
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
815.0001 815.0005 815.0010 815.0015 815.0020 815.0025 815.0030 815.0040 815.0050 815.0060 815.0070 815.0075 815.0080 815.0090 815.0100 815.0120 815.0150 815.0200 815.0250 815.0300 815.0400 815.0500 815.0600 815.0700 815.0750 815.0800 815.0900 815.1000 815.0010Y
SOCO815.0001EA 249 EUR
SOCO815.0001 SOCO815.0005 613-0559 SOCO815.0015 SOCO815.0020 SOCO815.0025 SOCO815.0030 SOCO815.0040 613-0560 SOCO815.0060 SOCO815.0070 SOCO815.0075 SOCO815.0080 SOCO815.0090 613-0561 SOCO815.0120 SOCO815.0150 613-0562 SOCO815.0250 SOCO815.0300 SOCO815.0400 613-0563 SOCO815.0600 SOCO815.0700 SOCO815.0750 SOCO815.0800 SOCO815.0900 613-0564 SOCO815.0010Y
Single channel micro pipettes, mechanical, fixed volume, Acura® manual 815
Pipetten Micropipetten
These pipettes show a very stable performance and provide consistent results for any analytical or routine tests.
  • Ergonomic shape, lightweight and soft plunger action
  • Volume setting is performed smoothly and precisely by turning push button; click-stops and free rotating caps prevent any unwanted volume alteration
  • Display is located at the front and therefore always visible at all times during pipetting
  • Justip™ tip ejector button
  • Autoclavable