Capillary micropipettes, variable volume, Acura® capillar 846
Leverancier: Socorex Isba
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
846.050 846.005 846.025 846.100 846.200
SOCO846.050EA 424 EUR
SOCO846.050 SOCO846.005 SOCO846.025 SOCO846.100 SOCO846.200
Capillary micropipettes, variable volume, Acura® capillar 846
Pipetten Micropipetten
High performance micropipettes with interchangeable glass capillary tube and ETFE tipped plunger.
  • Slim body shape, comfortable finger rest
  • Smooth plunger activation
  • Reliable, step-wise volume adjustment
  • Colour codes matching on pipettes, plungers and capillaries
  • Stainless steel, ETFE tipped plunger
The soft shaped handle and smooth plunger activation provide for enhanced hand comfort while handling viscous liquids, foaming solutions and volatile solvents.
Order Now
Specificaties
Environmentally Preferable