TriLink® CleanCap® Cas9 mRNA

Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L-7606-1000 L-7606-100 L-7606-5
SOLUL-7606-1000EA 1760 EUR
SOLUL-7606-1000 SOLUL-7606-100 SOLUL-7606-5
TriLink® CleanCap® Cas9 mRNA
Eiwitexpressie Reagentia voor eiwitexpressie

Cas9 mRNA expresses a version of the Streptococcus pyogenes SF370 Cas9 protein (CRISPR Associated Protein 9). Cas9 functions as part of the CRISPR (Clustered Regularly Interspaced Short Palindromic Repeats) genome editing system. In the CRISPR system, an RNA guide sequence targets the site of interest and the Cas9 protein is employed to perform double stranded DNA cleavage.


  • Capped with CleanCap® AG cap analog, no base modifications in the sequence

  • Cap 1 structure with >95% capping efficiency for superior in vivo activity compared to Cap-0 mRNAs

  • Manufactured with TriLink’s proprietary CleanScript® method to lower dsRNA for improved performance

  • Ready to use in downstream mRNA applications and formulations

  • Grade: RUO


This mRNA is capped using CleanCap® Reagent AG, TriLink's patented co-transcriptional capping technology, which results in the naturally occurring Cap-1 structure with >95% capping efficiency. It is manufactured using TriLink’s CleanScript® method, polyadenylated, and optimised for mammalian systems. It mimics a fully processed mature mRNA.



CleanCap Reagent AG produces a base-modified Cap-1 mRNA, which shows superior in vivo activity compared to Cap-0 mRNA produced by legacy capping methods such as mCap or anti-reverse cap analog (ARCA).



CleanScript method is TriLink’s proprietary in vitro transcription process that is optimised to minimise dsRNA formation and increase mRNA activity in vivo.



Opgelet: Item will be delivered at or below −40 °C and should be stored at or below −40 °C.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR