TriLink® 2'-Fluoro-2'-deoxyuridine-5'-Triphosphate
Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N-1010-1 N-1010-10 N-1010-100 N-1010-5
SOLUN-1010-1EA 62 EUR
SOLUN-1010-1 SOLUN-1010-10 SOLUN-1010-100 SOLUN-1010-5
TriLink® 2'-Fluoro-2'-deoxyuridine-5'-Triphosphate
Reagentia voor nucleïnezuren Nucleotiden
2'-Fluoro NTPs are being utilised in an increasing number of applications in research and new drug development. 2'-Fluoro-dUTP is incorporated in both DNA and RNA constructs to improve in vivo stability.
  • Grade: RUO
Both 2' Fluoro dU and dC are used in the design and synthesis of aptamers, antagomirs and siRNA, because they impart increased target affinity and resistance to endonuclease degradation, while reducing immune response. A key technique utilised in aptamer development using 2'-Fluoro-dUTP is SELEX (Systematic Evolution of Ligands by Exponential Enrichment). The SELEX process allows for the simultaneous screening of 1×1015 different oligonucleotides against a target of interest, such as a protein. The main goal of SELEX is to identify a small subset of aptamers from the original library that bind to the target of interest. Learn more in the technical article: An Overview of the SELEX Process.
Opgelet: Item will be delivered at or below −20 °C and should be stored at or below −20 °C.
New Product
Order Now
Specificaties
Environmentally Preferable
Applicatie