TriLink® 2'-Deoxy-P-nucleoside-5'-Triphosphate
Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
N-2037-1 N-2037-10 N-2037-5
SOLUN-2037-1EA 110 EUR
SOLUN-2037-1 SOLUN-2037-10 SOLUN-2037-5
TriLink® 2'-Deoxy-P-nucleoside-5'-Triphosphate
Reagentia voor nucleïnezuren Nucleotiden
2'-Deoxy-P-nucleoside is a degenerate base which behaves as a universal pyrimidine and therefore base pairs with both adenine and guanine. This feature makes it a powerful tool in mutagenesis studies.
  • Grade: RUO
Opgelet: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1fc7ca839d9b8b98eb9f5b6d1b3a8edaad080ec&vid=b1fc7ca839d9b8b98eb9f5b6d1b3a8edaad080ec&prid=web&clid=SAPDAM