TriLink® 2'-Deoxythymidine-5'-Triphosphate

Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N-2504-100 N-2504-25
SOLUN-2504-100EA 206 EUR
SOLUN-2504-100 SOLUN-2504-25
TriLink® 2'-Deoxythymidine-5'-Triphosphate
Reagentia voor nucleïnezuren Nucleotiden

This nucleotide consists of an unmodified base, a deoxyribose, and a 5′ triphosphate group for synthesis of DNA molecules.


  • Unmodified dexoyribonucleotide for DNA synthesis

  • ≥97% purity by AX-HPLC analysis

  • Provided as 100 mM in H₂O, pH 7,5

  • Grade: RUO


Deoxyadenosine-5′-triphosphate (dATP), deoxycytidine-5′-triphosphate (dCTP), deoxyguanosine-5′-triphosphate (dGTP), and deoxythymidine-5′-triphosphate (dTTP) are unmodified deoxyribonucleoside triphosphates, serving as essential building blocks of DNA molecules. Each of them consists of an unmodified base, a dexoyribose, and a 5′ triphosphate group.


Opgelet: Item will be delivered at or below −20 °C and should be stored at or below −20 °C.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d27446d02902a8ca92f05d8aaad4d196cd17e003&vid=d27446d02902a8ca92f05d8aaad4d196cd17e003&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR