TriLink® CleanCap® Reagent AG

Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N-7113-10 N-7113-1 N-7113-5 N-7113-100
SOLUN-7113-10EA 1070 EUR
SOLUN-7113-10 SOLUN-7113-1 SOLUN-7113-5 SOLUN-7113-100
TriLink® CleanCap® Reagent AG
Reagentia voor nucleïnezuren RNA Synthesis and Transcription

CleanCap® Reagent AG is TriLink’s patented co-transcriptional capping reagent for in vitro transcription of 5’ capped mRNA resulting in a Cap 1 structure. CleanCap has shown to provide up to 98% capping efficiency.


  • Grade: RUO


PLEASE NOTE: CleanCap Reagent AG requires an AG initiator. To learn more, please review the Product Insert.


Opgelet: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=658e4421b672768efb61f79ae1faca3a36a4c1d3&vid=658e4421b672768efb61f79ae1faca3a36a4c1d3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR