TriLink® CleanAmp® 7-deaza-dGTP Mix

Leverancier: TriLink BioTechnologies
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

N-9504-10 N-9504-2
SOLUN-9504-10EA 615 EUR
SOLUN-9504-10 SOLUN-9504-2
TriLink® CleanAmp® 7-deaza-dGTP Mix
Reagentia voor nucleïnezuren Nucleotiden

CleanAmp® dNTPs are chemically-modified nucleotides designed to improve specificity, sensitivity, and robustness of your PCR.


  • Improves PCR specificity, sensitivity, and robustness
  • Provides an alternative approach to hot-start PCR without a need for a specially designed DNA polymerase
  • Grade: RUO


The CleanAmp® 7-deaza-dGTP: - Contains CleanAmp® 7-deaza deoxyguanosine 5’ triphosphate in a 2 µmol or 10 µmol format - Is ideal for GC-rich PCR (~60% GC) due to promoting the template’s strand separation by lowering its melting temperature - Enables hot-start PCR for specificity and sensitivity, with a thermolabile protecting group at the 3’ hydroxyl of the nucleotides (CleanAmp® dNTP benefit) - Has been QC analysed by AX-HPLC, RP-HPLC, 31P NMR, mass spec, and a standardised PCR assay


Opgelet: Item will be delivered at or below –20 °C and should be stored at or below –20 °C.

https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5084b3b0e0412f89888e99c31de45dc5de5d06af&vid=5084b3b0e0412f89888e99c31de45dc5de5d06af&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR