Allantoin ≥98.5% USP

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A2721-25KGBL A2721-50KGBL A2721-500GM A2721-2.5KG
SPCMA2721-25KGBLEA 8090 EUR
SPCMA2721-25KGBL SPCMA2721-50KGBL SPCMA2721-500GM SPCMA2721-2.5KG
Allantoin ≥98.5% USP
Allantoin

Allantoin, USP is a moisturising ingredient.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formule: C₄H₆N₄O₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005260
CAS nummer: 97-59-6

Order Now

Specification Test Results

Assay 98.5 - 101.0%
Acidity or Alkalinity To Pass Test
Loss On Drying max. 0.1 %
Organic Impurities max. 0.5 %
Residue On Ignition (Sulphated Ash) max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR