Arachidyl alcohol

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A3850-5KGBL
SPCMA3850-5KGBLEA 0 EUR
SPCMA3850-5KGBL
Arachidyl alcohol
Arachidyl alcohol
Formule: H₃C(CH₂)₁₈CH₂OH
Kookpunt: 309 °C (1013 hPa)
Dichtheid: 0,88 g/cm³ (21 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002938
CAS nummer: 629-96-9
EINECS: 211-119-4

Order Now

Specification Test Results

Assay 95% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR