Calcium chloride dihydrate ≥99.0% FCC

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Calcium Chloride Dihydrate, Calcium dichloride

C1099-500GM
SPCMC1099-500GMEA 202 EUR
SPCMC1099-500GM
Calcium chloride dihydrate ≥99.0% FCC
Calcium chloride dihydrate

Calcium Chloride, Dihydrate, FCC is used as a sequestrant and firming agent in food products. It is considered as generally recognized as safe (GRAS) by the U.S. Food and Drug Administration. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities


GHS hazard : Exclamation mark

Formule: CaCl₂·2H₂O
MW: 147.01 g/mol
Kookpunt: 1600
Smeltpunt: 176
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149613
CAS nummer: 10035-04-8
Merck Index: 13,01660

Order Now

Specification Test Results

Assay 99.0 - 107.0 %
Arsenic (As) max. 3 mg/kg
Fluoride max. 0.004 %
Lead (Pb) max. 5 mg/kg
Magnesium and Alkali Salts max. 4.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR