Calcium pantothenate ≥97.0% FCC

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

C1143-100GM C1143-2.5KG
SPCMC1143-100GMEA 253 EUR
SPCMC1143-100GM SPCMC1143-2.5KG
Calcium pantothenate ≥97.0% FCC
Calcium pantothenate

Calcium Pantothenate, FCC is often used in dietary supplements because, as a salt, it is more stable than pantothenic acid (Vitamin B5). The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formule: C₁₈H₃₂CaN₂O₁₀
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002766
CAS nummer: 137-08-6

Order Now

Specification Test Results

Assay (Dried Basis) 97.0 - 103.0 %
Specific Rotation [A]D +25.0 to +27.5
Calcium Content 8.2 - 8.6 %
Loss On Drying max. 5.0 %
Alkalinity To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR