Calcium phosphate tribasic, powder NF

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Hydroxylapatite, tri-Calcium phosphate, HAp, Calcium Hydroxide Phosphate, Calcium hydroxyphosphate, Tribasic calcium phosphate

CA165-12KG CA165-500GM
SPCMCA165-12KGEA 1800 EUR
SPCMCA165-12KG SPCMCA165-500GM
Calcium phosphate tribasic, powder NF
Calcium phosphate tribasic

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formule: Ca₅HO₁₃P₃
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00010904
CAS nummer: 12167-74-7

Order Now

Specification Test Results

Assay (Ca) 34.0 - 40.0%
Loss On Ignition max. 8.0 %
Water-Soluble Substances max. 0.5 %
Acid-Insoluble Substances max. 0.2 %
Carbonate (CO₃) To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR