DL-Camphor (synthetic) USP

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

CA185-110LBBL CA185-2.5KG CA185-25KGBL
SPCMCA185-110LBBLEA 6830 EUR
SPCMCA185-110LBBL SPCMCA185-2.5KG SPCMCA185-25KGBL
DL-Camphor (synthetic) USP
DL-Camphor

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formule: C₁₀H₁₆O
Kookpunt: 204 °C (1013 hPa)
Smeltpunt: ∼180 °C
Dichtheid: 0,992 g/cm³ (25 °C)
Vlampunt: 66 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00074738
CAS nummer: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR