1,1,1-Trichloro-2-methyl-2-propanol, watervrij NF

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

CH123-12KGBL CH123-50KGBL CH123-25GM CH123-500GM
SPCMCH123-12KGBLEA 0 EUR
SPCMCH123-12KGBL SPCMCH123-50KGBL SPCMCH123-25GM SPCMCH123-500GM
1,1,1-Trichloro-2-methyl-2-propanol, watervrij NF
1,1,1-Trichloro-2-methyl-2-propanol

Melamine statement (available upon request)
BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formule: C₄H₇Cl₃O
Kookpunt: 166 °C (1013 hPa)
Smeltpunt: 78 °C
MDL Nummer: MFCD02179352
CAS nummer: 57-15-8
EINECS: 200-317-6
Merck Index: 13,02148

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR