2-(2-Ethoxyethoxy)ethyl acetate

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Diethylene glycol monoethyl ether acetate

E1023-200LTBL E1023-20LTMT
SPCME1023-200LTBLEA 5990 EUR
SPCME1023-200LTBL SPCME1023-20LTMT
2-(2-Ethoxyethoxy)ethyl acetate
2-(2-Ethoxyethoxy)ethyl acetate
Formule: C₈H₁₆O₄
MW: 176.21 g/mol
Kookpunt: 210
Smeltpunt: –25 °C
Dichtheid: 1.0114 g/cm³ (20 °C)
MDL Nummer: MFCD00041928
CAS nummer: 112-15-2
EINECS: 203-940-1

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR