Iron(II) sulphate heptahydrate ≥99.5%, granular FCC

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Iron vitriol, Ferrous sulphate heptahydrate

F1062-2.5KG F1062-500GM
SPCMF1062-2.5KGEA 548 EUR
SPCMF1062-2.5KG SPCMF1062-500GM
Iron(II) sulphate heptahydrate ≥99.5%, granular FCC
Ijzer(II)sulfaat heptahydraat

Ferrous Sulfate, Heptahydrate, Granular, FCC is used as an iron fortification food supplement additive. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark.

Formule: FeSO₄·7H₂O
MW: 278.02 g/mol
Smeltpunt: 64
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149719
CAS nummer: 7782-63-0
Merck Index: 13,04091

Order Now

Specification Test Results

Assay (FeSO₄.7H₂O) 99.5 - 104.5%
Lead (Pb) max. 2 mg/kg
Mercury max. 1 mg/kg
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR