Iron(II) fumarate ≥97.0% USP

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

FE108-12KG FE108-2.5KG FE108-50KGBL
SPCMFE108-12KGEA 1020 EUR
SPCMFE108-12KG SPCMFE108-2.5KG SPCMFE108-50KGBL
Iron(II) fumarate ≥97.0% USP
Iron(II) fumarate

Ferrous Fumarate, USP is a mineral supplement.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formule: C₄H₂FeO₄
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00058315
CAS nummer: 141-01-5

Order Now

Specification Test Results

Assay (C₄H₂Feo₄) 97.0-101.0%
Loss On Drying max.1.5 %
Sulfates (SO₄) max. 0.2 %
Arsenic (As) max. 3 ppm
Ferric Iron (Fe [D) max. 2.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR