Locust bean gum FCC

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

L1135-500GM L1135-250GM
SPCML1135-500GMEA 301 EUR
SPCML1135-500GM SPCML1135-250GM
Locust bean gum FCC
Locust bean gum

Locust (Carob) Bean Gum, FCC is a gelling and thickening agent used in certain foods. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00131257
CAS nummer: 9000-40-2

Order Now

Specification Test Results

Galactomannans 0.75
Acid-Insoluble Matter max.4.0 %
Arsenic (As) max. 3 mg/kg
Ash (Total) max. 1.2 %
Lead (Pb) max. 5 mg/kg


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR