L(+)-Lysine monohydrochloride, granular USP

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Lysine Hydrochloride, (S)-(+)-2,6-Diaminohexanoic acid monohydrochloride, L-Lysine monohydrochloride, H-Lys-OH·HCl, L-Lysine monohydrocholoride, L-Lysine Hydrochloride, (S)-(+)-Lysine monohydrochloride, (S)-(+)-2,6-Diaminocaproic acid monohydrochloride, H-Lys-OH.HCl

L1142-1KG L1142-50KGBL L1142-5KG
SPCML1142-1KGEA 346 EUR
SPCML1142-1KG SPCML1142-50KGBL SPCML1142-5KG
L(+)-Lysine monohydrochloride, granular USP
L(+)-Lysine monohydrochloride

Form: Granular
Use: excipient, biochemistry.

Formule: H₂N(CH₂)₄CH(NH₂)CO₂H·HCl
MW: 182.65 g/mol
Smeltpunt: 263…264 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00064564
CAS nummer: 657-27-2
EINECS: 211-519-9

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR