Papain
Leverancier: Spectrum Chemical
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
PA125-500GM PA125-100GM
SPCMPA125-500GMEA 869 EUR
SPCMPA125-500GM SPCMPA125-100GM
Papain
Enzymen
Papain, powder, purified, is a cysteine protease enzyme the is present in the papaya fruit.
It is traditionally used to break down meat fibres. It has more recently been used to dissociate cells in the early steps of cell culture preparation.