Riboflavine (vitamin B2) ≥98.0% FCC

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Vitamin B2, Lactoflavin, 7,8-Dimethyl-10-((2S,3S,4R)-2,3,4,5-tetrahydroxypentyl)benzo[g]pteridine-2,4(3H,10H)-dione, (-)-Riboflavin, Vitamine G

R1032-25GMGL
SPCMR1032-25GMGLEA 124 EUR
SPCMR1032-25GMGL
Riboflavine (vitamin B2) ≥98.0% FCC
Riboflavine (vitamin B2)

Riboflavin, FCC is also referred to as vitamin B2 and used as a dietary supplement. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formule: C₁₇H₂₀N₄O₆
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00005022
CAS nummer: 83-88-5

Order Now

Specification Test Results

Assay (Dried Basis) 98.0 - 102.0 %
Specific Rotation +56.5 to +59.5
Loss On Drying max. 1.5 %
Lumiflavin To Pass Test
Residue On Ignition max. 0.3 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR