Sodium fluoride ≥98.5%, powder USP, Ph. Eur., BP

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

SO167-125GM
SPCMSO167-125GMEA 138 EUR
SPCMSO167-125GM
Sodium fluoride ≥98.5%, powder USP, Ph. Eur., BP
Natriumfluoride

Sodium Fluoride, Powder, USP, EP, BP is used to prevent cavities making teeth stronger and more resistant to decay caused by acid and bacteria. Toothpaste being the preferred delivery method. All Spectrum Chemical USP, EP, BP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Skull and crossbones;Exclamation mark;Health hazard
UN1690

Formule: NaF
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003524
CAS nummer: 7681-49-4
UN: 1690
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2807d129a60e4d136eb782386d584317338c4499&vid=02396e9a739aa8e35f0b20961710e28c6ec76be0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (NaF; Dried Basis) 98.5 - 100.5%
Acidity or Alkalinity To Pass Test
Appearance of Solution To Pass Test
Loss On Drying max. 0.5%
Fluosilicate To Pass Test

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR