Sorbic acid, powder NF

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

SO215-12KG SO215-2.5KG SO215-100GM
SPCMSO215-12KGEA 1650 EUR
SPCMSO215-12KG SPCMSO215-2.5KG SPCMSO215-100GM
Sorbic acid, powder NF
Sorbic acid

BSE/TSE statement (available upon request)
Allergen statement (available upon request)

Formule: CH₃CH=CHCH=CHCOOH
Kookpunt: 228 °C (1013 hPa)
Smeltpunt: 132…135 °C
Dichtheid: 1,204 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00002703
CAS nummer: 110-44-1
EINECS: 203-768-7
Merck Index: 13,08793
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440c50b4fc52d9ff0b52afa7ba596c860f95e0e7&vid=f9b0ff409010208781696e9ca3314c02aaf1af36&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR