Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99% USP

Leverancier: Spectrum Chemical
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

Synoniemen: 2-Amino-2-(hydroxymethyl)propane-1,3-diol(tris), Tromethamine, Tris(hydroxymethyl)methylamine, 2-Amino-2-(hydroxymethyl)propane-1,3-diol, 2-Amino-2-(hydroxymethyl)-1,3-propanediol, Trizma(R) base, TRIS base, THAM

TR149-125GM TR149-1KG
SPCMTR149-125GMEA 102 EUR
SPCMTR149-125GM SPCMTR149-1KG
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) ≥99% USP
Tris-(hydroxymethyl)-aminomethaan (TRIS)

Tromethamine, USP is used as an emulsifying agent excipient.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formule: NH₂C(CH₂OH)₃
MW: 121.14 g/mol
Kookpunt: 357 °C (1013 hPa)
Smeltpunt: 169…172 °C
Dichtheid: 1,328 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00004679
CAS nummer: 77-86-1
EINECS: 201-064-4
Merck Index: 13,09842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=11a03d1ab6b2c11b9a2c83f0a73fc4264cec0110&vid=5ce890cdf298a59a7900a07fee1c857a5e5e61e0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR