Dodecamolybdophosphoric acid hydrate Microscopy, Supelco®

Leverancier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate

79560-100G
SUPL79560-100GEA 133 EUR
SUPL79560-100G
Dodecamolybdophosphoric acid hydrate Microscopy, Supelco®
Dodecamolybdophosphoric acid hydrate
Formule: H₃[P(Mo₃O₁₀)₄]·nH₂O
MW: 1843.27 g/mol
MDL Nummer: MFCD00149913
CAS nummer: 51429-74-4
Merck Index: 12,07498

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR