Resolvex® M10 Manual Positive Pressure Processor
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
288-0001 288-0006 289-0004
TCAT288-0001EA 0 EUR
TCAT288-0001 TCAT288-0006 TCAT289-0004
Resolvex® M10 Manual Positive Pressure Processor
Spruitstukken
Benchtop manual positive pressure processor.
  • Individual flow restrictors ensure parallel processing even when some wells are vacant
  • Only a gas supply is needed for operation
  • Dual-level gas delivery system offers fine controls of flow and pressure
Perfect for entry into positive pressure processing and workflows, the intuitive design of the Resolvex® M10 makes it easy to use with minimal training required.