SPE Plate, 96-well, NBE TRACE-N20
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
611-0051NBE
TCAT611-0051NBEEA 0 EUR
TCAT611-0051NBE
SPE Plate, 96-well, NBE TRACE-N20
Microplaten Platen voor vastefase-extractie
An end-capped C18 silica sorbent ideal for reverse-phase retention of neutral and polar analytes.
  • Reversed-phase
  • Liquid handler compatible
A 15 µm particulate with endcapped C18 chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for 25-OH-VitD/steroid assays.