SPE Plate, 96-well, NBE PSAX
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
615-0051NBE
TCAT615-0051NBEEA 0 EUR
TCAT615-0051NBE
SPE Plate, 96-well, NBE PSAX
Microplaten Platen voor vastefase-extractie
Mixed mode reverse-phase with an anion exchanger for acidic compounds.
  • Ion-exchange
  • Liquid handler compatible
A 17 µm polymer micro particulate with strong anionic exchange chemistry providing large contat zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Ideal for acidic target compounds.