SPE Column, NBE OFX
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
633-0031R-NBE
TCAT633-0031R-NBEEA 0 EUR
TCAT633-0031R-NBE
SPE Column, NBE OFX
Kolommen voor chromatografie
Proprietary blend of ion-exchange and water-wettable polymer targeted towards oral fluid collection devices.