Cerex® FNT SPE Column
Leverancier: Tecan
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
667-0101R
TCAT667-0101REA 0 EUR
TCAT667-0101R
Cerex® FNT SPE Column
Kolommen voor chromatografie
A water-wettable polymer with strong cation exchange chemistry.
  • Ion-exchange
  • Liquid handler compatible
A 15 µm micro particulate with strong cation exchange chemistry providing large contact zone for sorbent analyte interaction resulting in a fast and uniform sample prep. Formulated for FNT and pain panel.