N-Acetyl-β-D-glucosamine ≥98.0% (by HPLC, total nitrogen)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A0092-25G A0092-500G A0092-100G
TCIAA0092-25GEA 19.8 EUR
TCIAA0092-25G TCIAA0092-500G TCIAA0092-100G
N-Acetyl-β-D-glucosamine ≥98.0% (by HPLC, total nitrogen)
N-Acetyl-β-D-glucosamine
Formule: C₈H₁₅NO₆
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00136044
CAS nummer: 7512-17-6
EINECS: 231-368-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c84d4d1267d62db60de1a9764795c460ae962a47&vid=c84d4d1267d62db60de1a9764795c460ae962a47&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D +39.0 to +42.0 deg (C=1, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR