4-Acetylpyridine ≥98.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A0113-25G A0113-250G
TCIAA0113-25GEA 36.3 EUR
TCIAA0113-25G TCIAA0113-250G
4-Acetylpyridine ≥98.0% (by GC, titration analysis)
4-Acetylpyridine
Formule: C₇H₇NO
Kookpunt: 214°C
Smeltpunt: 14…16 °C
Dichtheid: 1,1009 g/cm³ (20 °C)
Vlampunt: 104°C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006433
CAS nummer: 1122-54-9
EINECS: 214-350-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=15b357eb4016faac725e17f27cff5daec9281a4f&vid=15b357eb4016faac725e17f27cff5daec9281a4f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR