Acetylthiocholine iodide ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

Synoniemen: (2-Mercaptoethyl)trimethylammonium iodide acetate, 2-(Acetylthio)-N,N,N-trimethylethan-1-aminium iodide, S-Acetylthiocholine iodide

A0116-25G A0116-1G A0116-5G
TCIAA0116-25GEA 321 EUR
TCIAA0116-25G TCIAA0116-1G TCIAA0116-5G
Acetylthiocholine iodide ≥98.0% (by titrimetric analysis)
Acetylthiocholine iodide
Formule: H₃CC(S)O(CH₂)₂N(CH₃)₃I
MW: 289.18 g/mol
Smeltpunt: 204
Bewaar temperatuur: Koelkast
MDL Nummer: MFCD00011819
CAS nummer: 1866-15-5
EINECS: 217-474-1
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=87d3f4d84d5d3b8bbe65be5d76730894b6566ecc&vid=87d3f4d84d5d3b8bbe65be5d76730894b6566ecc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR