Adenosine-5'-monophosphoric acid (AMP) ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A0158-1G A0158-25G A0158-5G
TCIAA0158-1GEA 17.6 EUR
TCIAA0158-1G TCIAA0158-25G TCIAA0158-5G
Adenosine-5'-monophosphoric acid (AMP) ≥98.0% (by HPLC, titration analysis)
Adenosine-5'-monophosphoric acid (AMP)
Formule: C₁₀H₁₄N₅O₇P
MDL Nummer: MFCD00005750
CAS nummer: 61-19-8
EINECS: 200-500-0
Merck Index: 14,00158
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e2c0f2dcc8f640b25fa3e4f1d390f6a2dc3b577&vid=8e2c0f2dcc8f640b25fa3e4f1d390f6a2dc3b577&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (after drying)
Specific rotation [a]20/D -45.0 to -50.0 deg (C=2, 1mol/L NaOH) (calcd.on dried substance)
Solubility in Sodium hydroxide solution almost transparency

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR