Allantoin ≥98.0% (by titrimetric analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

A0211-25G A0211-500G
TCIAA0211-25GEA 17.6 EUR
TCIAA0211-25G TCIAA0211-500G
Allantoin ≥98.0% (by titrimetric analysis)
Allantoin
Formule: C₄H₆N₄O₃
MDL Nummer: MFCD00005260
CAS nummer: 97-59-6
EINECS: 202-592-8
Merck Index: 14,00258
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=86fcc38b763cdc71466e55773f86fe29755a8530&vid=86fcc38b763cdc71466e55773f86fe29755a8530&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White powder to crystal
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR