Aniline-2,5-disulphonic acid monosodium salt ≥98.0% (by HPLC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A0263-25G A0263-500G
TCIAA0263-25GEA 23.1 EUR
TCIAA0263-25G TCIAA0263-500G
Aniline-2,5-disulphonic acid monosodium salt ≥98.0% (by HPLC, titration analysis)
Aniline-2,5-disulphonic acid monosodium salt
Formule: C₆H₇NO₆S₂
MDL Nummer: MFCD00078393
CAS nummer: 24605-36-5
EINECS: 246-349-4

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR