2-Aminofenol ≥99.0% (by GC)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

A0335-25G A0335-500G A0335-100G
TCIAA0335-25GEA 50.6 EUR
TCIAA0335-25G TCIAA0335-500G TCIAA0335-100G
2-Aminofenol ≥99.0% (by GC)
2-Aminofenol
Formule: C₆H₇NO
Kookpunt: 235 °C (1013 hPa)
Dichtheid: 1,328 g/cm³ (20 °C)
Vlampunt: ≥ 175 °C
MDL Nummer: MFCD00007690
CAS nummer: 95-55-6
EINECS: 202-431-1
UN: 2512
ADR: 6.1,III
Merck Index: 14,00461
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c1a634727cccba28855fb83c45b6d032ee3797b1&vid=c1a634727cccba28855fb83c45b6d032ee3797b1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Solubility in dilute HCl within almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR