3-Aminopyridine ≥99.0% (by GC, titration analysis)

Leverancier: TCI
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

A0413-25G A0413-500G A0413-100G
TCIAA0413-25GEA 23.1 EUR
TCIAA0413-25G TCIAA0413-500G TCIAA0413-100G
3-Aminopyridine ≥99.0% (by GC, titration analysis)
3-Aminopyridine
Formule: C₅H₆N₂
Kookpunt: 248°C
Smeltpunt: 60…63 °C
Vlampunt: 88 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00006400
CAS nummer: 462-08-8
EINECS: 207-322-2
UN: 2671
ADR: 6.1,II
Merck Index: 14,00473
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93dba9e6da98ade269a0d230bf656616f7295555&vid=93dba9e6da98ade269a0d230bf656616f7295555&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 99.0 %
Purity (Nonaqueous Titration) min. 99.0 %
Melting point 59.0 to 65.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR